N1-Benzyl-N1-methyl-1,3-propanediamine

Supplier: Matrix Scientific

026894-500MG
101823-084EA 306.15 USD
101823-084
N1-Benzyl-N1-methyl-1,3-propanediamine
N1-Benzyl-N1-methyl-1,3-propanediamine

Hazard: Irritant

Formula: C₁₁H₁₈N₂
MDL Number: MFCD03001135
CAS Number: 13910-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR