N1-(2-Furylmethyl)-1,3-propanediamine

Supplier: Matrix Scientific

026893-500MG
101823-082EA 306.15 USD
101823-082
N1-(2-Furylmethyl)-1,3-propanediamine
N1-(2-Furylmethyl)-1,3-propanediamine

Hazard: Irritant

Formula: C₈H₁₄N₂O
MDL Number: MFCD06213178
CAS Number: 21878-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR