3-[(2-Furylmethyl)amino]propanenitrile

Supplier: Matrix Scientific

026892-500MG
101823-080EA 203.08 USD
101823-080
3-[(2-Furylmethyl)amino]propanenitrile
3-[(2-Furylmethyl)amino]propanenitrile

Hazard: Irritant

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
MDL Number: MFCD00219840
CAS Number: 6788-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR