Ethyl-3-[(2-furylmethyl)amino]propanoate

Supplier: Matrix Scientific

026890-500MG
101823-076EA 306.15 USD
101823-076
Ethyl-3-[(2-furylmethyl)amino]propanoate
Ethyl-3-[(2-furylmethyl)amino]propanoate

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
MW: 197.24 g/mol
MDL Number: MFCD00219838
CAS Number: 175203-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR