Methyl-3-[(2-furylmethyl)amino]propanoate

Supplier: Matrix Scientific

026889-500MG
101823-074EA 306.15 USD
101823-074
Methyl-3-[(2-furylmethyl)amino]propanoate
Methyl-3-[(2-furylmethyl)amino]propanoate

Hazard: Irritant

Formula: C₉H₁₃NO₃
MDL Number: MFCD01475841
CAS Number: 4063-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR