N1,N1-Dipropyl-1,3-propanediamine

Supplier: Matrix Scientific

026887-500MG
101823-070EA 495.38 USD
101823-070
N1,N1-Dipropyl-1,3-propanediamine
N1,N1-Dipropyl-1,3-propanediamine

Hazard: Irritant

Formula: C₉H₂₂N₂
MDL Number: MFCD00014830
CAS Number: 6345-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR