3-[Cyclohexyl(methyl)amino]propanohydrazide

Supplier: Matrix Scientific

026886-500MG
101823-068EA 306.15 USD
101823-068
3-[Cyclohexyl(methyl)amino]propanohydrazide
3-[Cyclohexyl(methyl)amino]propanohydrazide

Hazard: Irritant

Formula: C₁₀H₂₁N₃O
MDL Number: MFCD10687296
CAS Number: 1040689-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR