N1-Cyclohexyl-N1-methyl-1,3-propanediamine

026885-500MG
101823-066EA 1988.46 USD
101823-066
N1-Cyclohexyl-N1-methyl-1,3-propanediamine
N1-Cyclohexyl-N1-methyl-1,3-propanediamine

Hazard: Irritant

Formula: C₁₀H₂₂N₂
MDL Number: MFCD06446874
CAS Number: 26735-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR