N-Isopropyl-1,3-propanediamine

Supplier: Matrix Scientific

Synonyms: N-Isopropyl-1,3-propanediamineN-Isopropyl-1,3-diaminopropane3-(Isopropylamino)propylamine

026882-500MG
101823-060EA 306.15 USD
101823-060
N-Isopropyl-1,3-propanediamine
N-Isopropyl-1,3-propanediamine

Hazard: Irritant

Formula: C₆H₁₆N₂
MDL Number: MFCD00008208
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR