Ethyl-3-(isopropylamino)propanoate

Supplier: Matrix Scientific

026879-500MG
101823-054EA 306.15 USD
101823-054
Ethyl-3-(isopropylamino)propanoate
Ethyl-3-(isopropylamino)propanoate

Hazard: Irritant

Formula: C₈H₁₇NO₂
MDL Number: MFCD01475850
CAS Number: 16217-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR