Methyl-3-(isopropylamino)propanoate

Supplier: Matrix Scientific

026878-500MG
101823-052EA 306.15 USD
101823-052
Methyl-3-(isopropylamino)propanoate
Methyl-3-(isopropylamino)propanoate

Hazard: Irritant

Formula: C₇H₁₅NO₂
MDL Number: MFCD01475814
CAS Number: 42313-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR