3-(4-Methyl-1-piperazinyl)propanohydrazide

Supplier: Matrix Scientific

026877-500MG
101823-050EA 306.15 USD
101823-050
3-(4-Methyl-1-piperazinyl)propanohydrazide
3-(4-Methyl-1-piperazinyl)propanohydrazide

Hazard: Irritant

Formula: C₈H₁₈N₄O
MDL Number: MFCD04624246
CAS Number: 24636-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR