3-(Cyclohexylamino)propanamide
Supplier: Matrix Scientific
026874-500MG
101823-044EA
306.15
USD
101823-044
3-(Cyclohexylamino)propanamide
3-(Cyclohexylamino)propanamide
Hazard: Irritant
Formula:
C₉H₁₈N₂O |
MDL Number:
MFCD01475860 CAS Number: 4552-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...