Ethyl-3-(cyclohexylamino)propanoate

Supplier: Matrix Scientific

026873-500MG
101823-042EA 306.15 USD
101823-042
Ethyl-3-(cyclohexylamino)propanoate
Ethyl-3-(cyclohexylamino)propanoate

Hazard: Irritant

Formula: C₁₁H₂₁NO₂
MDL Number: MFCD01475857
CAS Number: 6635-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR