Methyl-3-(cyclohexylamino)propanoate

Supplier: Matrix Scientific

026871-500MG
101823-038EA 306.15 USD
101823-038
Methyl-3-(cyclohexylamino)propanoate
Methyl-3-(cyclohexylamino)propanoate

Hazard: Irritant

Formula: C₁₀H₁₉NO₂
MDL Number: MFCD00482233
CAS Number: 22870-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR