3-[(Tetrahydro-2-furanylmethyl)amino]propanohydrazide

Supplier: Matrix Scientific

026865-500MG
101823-026EA 306.15 USD
101823-026
3-[(Tetrahydro-2-furanylmethyl)amino]propanohydrazide
3-[(Tetrahydro-2-furanylmethyl)amino]propanohydrazide

Hazard: Irritant

Formula: C₈H₁₇N₃O₂
MW: 187.24 g/mol
MDL Number: MFCD10687287
CAS Number: 1040688-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR