N1-(Tetrahydro-2-furanylmethyl)-1,3-propanediamine

Supplier: Matrix Scientific

026864-500MG
101823-024EA 306.15 USD
101823-024
N1-(Tetrahydro-2-furanylmethyl)-1,3-propanediamine
N1-(Tetrahydro-2-furanylmethyl)-1,3-propanediamine

Hazard: Irritant

Formula: C₈H₁₈N₂O
MDL Number: MFCD06213150
CAS Number: 90203-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR