3-(Hexadecylamino)propanenitrile

Supplier: Matrix Scientific

026861-500MG
101823-018EA 306.15 USD
101823-018
3-(Hexadecylamino)propanenitrile
3-(Hexadecylamino)propanenitrile

Hazard: Irritant

Formula: C₁₉H₃₈N₂
MDL Number: MFCD10687285
CAS Number: 36341-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR