Methyl-3-[(tetrahydro-2-furanylmethyl)amino]propanoate

Supplier: Matrix Scientific

026859-500MG
101823-014EA 306.15 USD
101823-014
Methyl-3-[(tetrahydro-2-furanylmethyl)amino]propanoate
Methyl-3-[(tetrahydro-2-furanylmethyl)amino]propanoate

Hazard: Irritant

Formula: C₉H₁₇NO₃
MDL Number: MFCD10687283
CAS Number: 1040688-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR