3-(1-Pyrrolidinyl)propanohydrazide

Supplier: Matrix Scientific

026858-500MG
101823-012EA 306.15 USD
101823-012
3-(1-Pyrrolidinyl)propanohydrazide
3-(1-Pyrrolidinyl)propanohydrazide

Hazard: Irritant

Formula: C₇H₁₅N₃O
MDL Number: MFCD01839708
CAS Number: 59737-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR