3-(Heptylamino)propanenitrile

Supplier: Matrix Scientific

026854-500MG
101823-004EA 306.15 USD
101823-004
3-(Heptylamino)propanenitrile
3-(Heptylamino)propanenitrile

Hazard: Irritant

Formula: C₁₀H₂₀N₂
MDL Number: MFCD10687280
CAS Number: 101421-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR