Methyl-3-(heptylamino)propanoate

Supplier: Matrix Scientific

026851-500MG
101822-998EA 306.15 USD
101822-998
Methyl-3-(heptylamino)propanoate
Methyl-3-(heptylamino)propanoate

Hazard: Irritant

Formula: C₁₁H₂₃NO₂
MDL Number: MFCD10687277
CAS Number: 40870-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR