3-(Hexadecylamino)propanamide

Supplier: Matrix Scientific

026850-500MG
101822-996EA 306.15 USD
101822-996
3-(Hexadecylamino)propanamide
3-(Hexadecylamino)propanamide

Hazard: Irritant

Formula: C₁₉H₄₀N₂O
MDL Number: MFCD10687276
CAS Number: 36341-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR