N1-(1-Phenylethyl)-1,3-propanediamine

Supplier: Matrix Scientific

026847-500MG
101822-990EA 306.15 USD
101822-990
N1-(1-Phenylethyl)-1,3-propanediamine
N1-(1-Phenylethyl)-1,3-propanediamine

Hazard: Irritant

Formula: C₁₁H₁₈N₂
MDL Number: MFCD03596779
CAS Number: 52198-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR