3-[(1-Phenylethyl)amino]propanenitrile

Supplier: Matrix Scientific

026846-500MG
101822-988EA 306.15 USD
101822-988
3-[(1-Phenylethyl)amino]propanenitrile
3-[(1-Phenylethyl)amino]propanenitrile

Hazard: Irritant

Formula: C₁₁H₁₄N₂
MDL Number: MFCD08234521
CAS Number: 112971-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR