Ethyl-3-[(1-phenylethyl)amino]propanoate

Supplier: Matrix Scientific

026844-500MG
101822-984EA 306.15 USD
101822-984
Ethyl-3-[(1-phenylethyl)amino]propanoate
Ethyl-3-[(1-phenylethyl)amino]propanoate

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD00480752
CAS Number: 59576-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR