Methyl-3-[(1-phenylethyl)amino]propanoate
Supplier: Matrix Scientific
026843-500MG
101822-982EA
306.15
USD
101822-982
Methyl-3-[(1-phenylethyl)amino]propanoate
Methyl-3-[(1-phenylethyl)amino]propanoate
Hazard: Irritant
Formula:
C₁₂H₁₇NO₂ |
MDL Number:
MFCD00540146 CAS Number: 42792-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...