3-(tert-Butylamino)propanamide

Supplier: Matrix Scientific

026840-500MG
101822-976EA 306.15 USD
101822-976
3-(tert-Butylamino)propanamide
3-(tert-Butylamino)propanamide

Hazard: Irritant

Formula: C₇H₁₆N₂O
MDL Number: MFCD01475862
CAS Number: 289656-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR