Ethyl-3-(tert-butylamino)propanoate

Supplier: Matrix Scientific

026838-500MG
101822-972EA 306.15 USD
101822-972
Ethyl-3-(tert-butylamino)propanoate
Ethyl-3-(tert-butylamino)propanoate

Hazard: Irritant

Formula: C₉H₁₉NO₂
MDL Number: MFCD01475854
CAS Number: 1462-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR