Methyl-3-(tert-butylamino)propanoate

Supplier: Matrix Scientific

026837-500MG
101822-970EA 306.15 USD
101822-970
Methyl-3-(tert-butylamino)propanoate
Methyl-3-(tert-butylamino)propanoate

Hazard: Irritant

Formula: C₈H₁₇NO₂
MDL Number: MFCD01475840
CAS Number: 21539-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR