[1-(3-Methoxypropyl)-3-pyrrolidinyl]methanamine

Supplier: Matrix Scientific

026831-500MG
101822-958EA 203.08 USD
101822-958
[1-(3-Methoxypropyl)-3-pyrrolidinyl]methanamine
[1-(3-Methoxypropyl)-3-pyrrolidinyl]methanamine

Hazard: Irritant

Formula: C₉H₂₀N₂O
MDL Number: MFCD09759248
CAS Number: 914202-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR