3,5-Dimethyl-1,2-benzenediamine

Supplier: Matrix Scientific

026828-500MG
101822-952EA 210.77 USD
101822-952
3,5-Dimethyl-1,2-benzenediamine
3,5-Dimethyl-1,2-benzenediamine

Hazard: Irritant

Formula: C₈H₁₂N₂
MDL Number: MFCD00972722
CAS Number: 3171-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR