1-Phenyl-1-propanone oxime

Supplier: Matrix Scientific

026826-500MG
101822-948EA 203.08 USD
101822-948
1-Phenyl-1-propanone oxime
1-Phenyl-1-propanone oxime

Hazard: Irritant

Formula: C₉H₁₁NO
MDL Number: MFCD01197334
CAS Number: 23517-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR