2-Ethoxy-3-methoxybenzaldehyde oxime

Supplier: Matrix Scientific

026825-500MG
101822-946EA 306.15 USD
101822-946
2-Ethoxy-3-methoxybenzaldehyde oxime
2-Ethoxy-3-methoxybenzaldehyde oxime

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD09703914
CAS Number: 80364-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR