3-(Benzylamino)propionitrile

Supplier: Matrix Scientific

Synonyms: 3-(Benzylamino)propanenitrile

026823-500MG
101822-942EA 306.15 USD
101822-942
3-(Benzylamino)propionitrile
3-(Benzylamino)propionitrile

Hazard: Irritant

Formula: C₁₀H₁₂N₂
MW: 160.22 g/mol
MDL Number: MFCD00001955
CAS Number: 706-03-6
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR