1-(4-Ethoxyphenyl)-1-propanone oxime

Supplier: Matrix Scientific

026822-500MG
101822-940EA 306.15 USD
101822-940
1-(4-Ethoxyphenyl)-1-propanone oxime
1-(4-Ethoxyphenyl)-1-propanone oxime

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD09945739
CAS Number: 91247-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR