3-Methoxy-2-propoxybenzaldehyde oxime

Supplier: Matrix Scientific

026819-500MG
101822-934EA 306.15 USD
101822-934
3-Methoxy-2-propoxybenzaldehyde oxime
3-Methoxy-2-propoxybenzaldehyde oxime

Hazard: Irritant

Formula: C₁₁H₁₅NO₃
MDL Number: MFCD09710937
CAS Number: 41828-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR