N-Cycloheptyl-N-ethylamine

Supplier: Matrix Scientific

026817-500MG
101822-930EA 203.08 USD
101822-930
N-Cycloheptyl-N-ethylamine
N-Cycloheptyl-N-ethylamine

Hazard: Irritant

Formula: C₉H₁₉N
MDL Number: MFCD08691721
CAS Number: 45806-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR