1-(4-Ethoxyphenyl)-1-propanol

Supplier: Matrix Scientific

026815-500MG
101822-926EA 692.31 USD
101822-926
1-(4-Ethoxyphenyl)-1-propanol
1-(4-Ethoxyphenyl)-1-propanol

Hazard: Irritant

Formula: C₁₁H₁₆O₂
MDL Number: MFCD09991469
CAS Number: 1031927-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR