(4-Isopropoxy-3-methoxyphenyl)methanamine hydrochloride

Supplier: Matrix Scientific

026813-500MG
101822-922EA 181.54 USD
101822-922
(4-Isopropoxy-3-methoxyphenyl)methanamine hydrochloride
(4-Isopropoxy-3-methoxyphenyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClNO₂
MDL Number: MFCD09991467
CAS Number: 854185-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR