3-sec-Butoxybenzylamine

Supplier: Matrix Scientific

026810-500MG
101822-916EA 306.15 USD
101822-916
3-sec-Butoxybenzylamine
3-sec-Butoxybenzylamine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD09991465
CAS Number: 37806-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR