2-Propoxybenzylamine

Supplier: Matrix Scientific

026809-500MG
101822-914EA 203.08 USD
101822-914
2-Propoxybenzylamine
2-Propoxybenzylamine

Hazard: Irritant

Formula: C₁₀H₁₅NO
MDL Number: MFCD06212925
CAS Number: 37806-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR