3-[(3-Pyridinylmethyl)amino]propanohydrazide

Supplier: Matrix Scientific

026798-500MG
101822-892EA 306.15 USD
101822-892
3-[(3-Pyridinylmethyl)amino]propanohydrazide
3-[(3-Pyridinylmethyl)amino]propanohydrazide

Hazard: Irritant

Formula: C₉H₁₄N₄O
MDL Number: MFCD10687260
CAS Number: 1040688-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR