Ethyl-3-[(3-pyridinylmethyl)amino]propanoate

Supplier: Matrix Scientific

026794-500MG
101822-884EA 306.15 USD
101822-884
Ethyl-3-[(3-pyridinylmethyl)amino]propanoate
Ethyl-3-[(3-pyridinylmethyl)amino]propanoate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD10687258
CAS Number: 1040688-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR