3-(Diethylamino)propanohydrazide

Supplier: Matrix Scientific

026790-500MG
101822-876EA 495.38 USD
101822-876
3-(Diethylamino)propanohydrazide
3-(Diethylamino)propanohydrazide

Hazard: Irritant

Formula: C₇H₁₇N₃O
MDL Number: MFCD05267739
CAS Number: 83742-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR