3-[(2-Chlorobenzyl)amino]propanamide

Supplier: Matrix Scientific

026789-500MG
101822-874EA 306.15 USD
101822-874
3-[(2-Chlorobenzyl)amino]propanamide
3-[(2-Chlorobenzyl)amino]propanamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD10687254
CAS Number: 802262-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR