Ethyl-3-[(2-chlorobenzyl)amino]propanoate

Supplier: Matrix Scientific

026788-500MG
101822-872EA 306.15 USD
101822-872
Ethyl-3-[(2-chlorobenzyl)amino]propanoate
Ethyl-3-[(2-chlorobenzyl)amino]propanoate

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₂
MDL Number: MFCD03450398
CAS Number: 67044-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR