3-[(3-Methoxypropyl)amino]propanohydrazide

Supplier: Matrix Scientific

026786-500MG
101822-868EA 306.15 USD
101822-868
3-[(3-Methoxypropyl)amino]propanohydrazide
3-[(3-Methoxypropyl)amino]propanohydrazide

Hazard: Irritant

Formula: C₇H₁₇N₃O₂
MDL Number: MFCD10687252
CAS Number: 1040687-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR