N1-(3-Methoxypropyl)-1,3-propanediamine

Supplier: Matrix Scientific

026785-500MG
101822-866EA 495.38 USD
101822-866
N1-(3-Methoxypropyl)-1,3-propanediamine
N1-(3-Methoxypropyl)-1,3-propanediamine

Hazard: Irritant/Stored under Argon

Formula: C₇H₁₈N₂O
MDL Number: MFCD10687251
CAS Number: 68123-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR