3-[(3-Methoxypropyl)amino]propanenitrile

Supplier: Matrix Scientific

026784-500MG
101822-864EA 306.15 USD
101822-864
3-[(3-Methoxypropyl)amino]propanenitrile
3-[(3-Methoxypropyl)amino]propanenitrile

Hazard: Irritant

Formula: C₇H₁₄N₂O
MDL Number: MFCD10687250
CAS Number: 1040687-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR